Description
- The long handled spoons mean it is easy to scoop powder and liquid from a distance.
- It also means they can be easily seen if left in the powder.
- These single use spoons are manufactured in a cleanroom and packed in a class 7 cleanroom to ensure cleanliness.
- They are all individually bagged and so can be used straight from the bag without the need for costly cleaning.
- The spoons are available in a straight or an angled shape.
- The sample spoons are ideal for both sampling and dispensing powders, granules and liquids.
- The Single Use spoons comes pre bagged and ready to use.
- Simply unwrap and use.
- No cleaning or cleaning validation is required.
- They are available in 2 designs.
- The straight design is the most popular, whilst the ergonomic angled design means it is easier for right handled people to operate.
- Made from food grade HDPE.
- Conforms to the latest FDA and EU requirements.
- BSE/TSE free.
- HDPE is ideal for most applications as it is strong and rigid.
- The sterile spoons come with a certificate of irradiation.
- Range of sizes with different angled and straight combinations.
Specifications
Specifications
| Sterility | Non-sterile |
| Shape | Straight |
| Capacity (Metric) | 10 mL |
| Material | HDPE |