missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ SteriWare™ Disposable PowderSafe
BIOPHARMA

Product Code. 11975846 Shop All Sampling Systems Products
Change view
Click to view available options
Sterility:
Non-sterile
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility unitSize
11975846 Non-sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11975846 Supplier Sampling Systems™ Supplier No. 8097A150
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Powder Spatula complete with a see through, click-on container. The container slips over the blade of the spatula and is securely locked into place. At the laboratory the container can be easily removed.

Key Features:

  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Supplied with see-through container that securely clips over the spatula

Why Use a SteriWare™ Disposable Sampler?

  • No Cross Contamination - All SteriWare™ devices can be used straight from the bag.
  • No Cleaning - Unlike a reusable device, you can use a SteriWare™ product straight away. This eliminates the need for costly Cleaning Validation.
  • High Quality - Our SteriWare™ products are manufactured/moulded/packed in a Class 100,000 Cleanroom in England. Please see the products Certificate of Conformity and Technical Specification Sheet for more information.
  • Full Documentation - We can provide full batch specific documentation for all SteriWare™ products. This includes Certificates of Conformity and Certificates of Irradiation (where applicable). The batch number is clearly printed on every bag and box.
  • Cost Effective - Removing the need for Cleaning Validation by switching to a SteriWare™ device saves time and money.
  • Available Sterile - Most SteriWare™ products are available pre-sterilized by Gamma or Electron Beam to SAL10-6. Please see the Specification Sheet for more information.

Specifications

Sterility Non-sterile
Color White Spatula, Transparent Cover
Certifications/Compliance Conforms to FDA CFR 177.1640, Conforms to EU Regulation 10/2011
Disposable Single-use
Product Type Powder Sampler
For Use With (Application) For Easy, Controlled Powder Discharge
Material Polystyrene
Length (Metric) 230 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.