missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ SteriWare™ Disposable Knife
BIOPHARMA

Product Code. 15561960 Shop All Sampling Systems Products
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 15561960

Brand: Sampling Systems™ 8039H100S

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use straight from the bag without the need for costly cleaning. Sampling Systems™ SteriWare™ Disposable Knife is ideal for preparing samples as well as general laboratory use.

  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No Cross Contamination
  • No Cleaning required

Specifications

Sterility Sterile
Color White
Disposable Single-use
Length (Metric) Handle 100 mm
Material HDPE
Length (Metric) 160 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.