Learn More
Sampling Systems™ SteriWare™ Blue Disposable PharmaScoop™
GREENER_CHOICEBIOPHARMA
Product Code. 16691123 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 85
Product Code. 16691123
Brand: Sampling Systems™ 8045B250
Only null left
Add to basket
Add to basket
Use straight from the bag without the need for costly cleaning. Sampling Systems™ SteriWare™ Blue Disposable PharmaScoop™ is ideal for both sampling and dispensing powders and granules.
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged in ultra-strong packing with easy to open design
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Large variety of different sizes available
  • Good Manufacturing Practice (GMP) to keep your hands out of the product as much as possible
  • Positioned the ergonomic handle high up on the scoop
  • No Cross Contamination
  • No Cleaning required
Specifications
Sterility Non-sterile
Capacity (Metric) 250 mL
Color Blue
Disposable Single-use
Product Type Disposable Scoop
Includes Individually Bagged
Material Polypropylene