Learn More
Sampling Systems™ SteriWare™ Blue Disposable PharmaScoop™
GREENER_CHOICE
Product Code. 15839431 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 85
Product Code. 15839431
Brand: Sampling Systems™ 8045B250
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use straight from the bag without the need for costly cleaning. Sampling Systems™ SteriWare™ Blue Disposable PharmaScoop™ is ideal for both sampling and dispensing powders and granules.
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged in ultra-strong packing with easy to open design
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Large variety of different sizes available
  • Good Manufacturing Practice (GMP) to keep your hands out of the product as much as possible
  • Positioned the ergonomic handle high up on the scoop
  • No Cross Contamination
  • No Cleaning required
Specifications
Sterility Non-sterile
Capacity (Metric) 250 mL
Color Blue
Disposable Single-use
Product Type Disposable Scoop
Includes Individually Bagged
Material Polypropylene