missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ SteriPlast™ Bio Sample Scoops

Stands horizontally so the scoop can be put down without the sample material trickling out. Buerkle™ SteriPlast™ Bio Sample Scoops are designed with a slightly angled front edge, and are completely recyclable.

Brand:  Buerkle™ 5379-1003

Code : 03

Additional Details : Weight : 0.21300kg

 View more versions of this product

Product Code. 15346337

  • £22.00 / Pack of 10


Estimated Shipment
05-02-2019
Add to basket

Description

Description

  • Ergonimically designed scoop
  • Product and packaging made of renewable raw materials
  • Manufactured from "green" PE, white
  • Recyclable/Disposable
  • Individually packaged and sterilised by gamma rays
  • Production and packaging according to clean room class 7
  • With EU foodstuffs and FDA approval
Specifications

Specifications

White
160mm
Sterile
Scoop
Ergonomic
Green Polyethylene (PE made from renewable raw materials)
50mL
Videos
Documentation

Technical Documents