missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sterile PES Syringe Filter

Product Code. 15206869 Shop All Fisherbrand Products
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
15206869 0.2 μm Pack of 50
15216869 0.45 μm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15206869
Add to basket
Edge
Add to basket

Designed for filtration and purification in a sterile environment. Fisherbrand™ Sterile PES Syringe Filter features a PES membrane in a polypropylene housing for purification of tissue culture solutions from a standard syringe. 33mm diameter membranes available with 0.2 or 0.45μm pore sizes.

  • Ideal for purification of tissue culture solutions and buffers
  • High sample throughput with low protein binding
  • Hydrophilic 33mm polyethersulfone (PES) membrane
  • Sturdy, chemically resistant polypropylene housing

Specifications

Color White
Material (Membrane) PES
Pore Size 0.2 μm
Sterility Sterile
For Use With (Application) Purification of tissue culture solutions, buffers
Housing Material Polypropylene
Quantity 50/Pk.
Type Syringe Filter
Diameter (Metric) 33 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.