missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microspec™ Sterile Cribbar Sample Bottles (Volume: 1000mL)
missing translation for 'orderingAttributeHoverText'
Cap Color:
Blue
Green
White
Concentration:
10mg/L when full
10mg/L, undosed
18 mg/L
180mg/L, undosed
18mg/L when full
18mg/L, undosed
Additive:
EDTA
None
Sdium Thiosulphate
Sodium Thiosulphate
Color:
Green
Translucent
Förpackningsstorlek
Pack of 56
Beskrivelse
- For sampling, potability testing, Legionella spp, pathogenic and indicator species
- Squat bottles facilitating easy sampling from low drain points
- Features wide diameter necks for ease of sampling and pipetting
- Caps are tamper evident
- Stable on bench
- Rectangular to make most of limited storage space
- Squat Bottle for ease of sampling
- Wide neck for ease of sampling and pipetting
- Off-centre neck to aid sampling close fitting outlets
- Includes label for marking
Tekniske data
Tekniske data
| ATCC® | Yes |
| Cap Color | White |
| Cap Type | Screw |
| Color | Green |
| Concentration | 18mg/L, undosed |
| Content And Storage | Room Temperature |
| Additive | Sdium Thiosulphate |
| Detects | Microorganisms |
| Format | Disposable |
| Manufacturing Origin | United Kingdom |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion