missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ StemPro™ Neural Stem Cells
Description
StemPro™ Neural Stem Cells are cryopreserved human fetal brain-derived neural stem cells (NSCs). These cells are manufactured under Good Manufacturing Practice (GMP) manufacturing standards in a California-licensed facility and are available for Research Use Only. They are isolated and expanded in culture to passage 4 under a low oxygen (ischemic tolerant,) proprietary manufacturing process using FBS-supplemented media and then cryopreserved in an animal origin-free cryopreservation medium. A Certificate of Analysis is available for each lot that documents the quality control specifications, test results, and donor information. These cells have no observed tumorigenicity/toxicity in GLP-compliant animal studies.
StemPro™ Neural Stem Cells help you to:
Minimize assay variability with itNSCs manufactured using GMP manufacturing standards
Improve functionality studies with immature, high potency NSCs
Complete experiments requiring one large lot of NSCs
Minimize your chance of assay failure by using tested cells, reagents, and protocols
Immature, High Potency, Low Oxygen-Cultured NSCs
StemPro™ Neural Stem Cells are derived from human fetal brain from qualified, traceable donors. The cells are isolated, cultured, and expanded under Good Manufacturing Practice (GMP) manufacturing standards in a California-licensed facility using a proprietary Reduced Oxygen Tension manufacturing process. Manufacture of cells in a reduced oxygen tension environment results in higher yields of highly potent immature stem cells compared to cells expanded in normal oxygen culture conditions.
Safety Tested
StemPro™ Neural Stem Cells have been demonstrated to be non-toxic and non-tumorigenic in GLP animal studies performed according to FDA guidelines.
Large lots can be made available for those requiring assay standardization. Protocols using tested reagents are provided to help minimize your chance of assay failure using reagents and cells that have not been evaluated to work together. View our tested media and reagents.
High Post-Cryopreservation Viability
These StemPro™ Neural Stem Cells are supplied in CryoStor™ cryopreservation medium at a concentration of 5x106 viable cells/vial. CryoStor™ medium is animal origin-free and shown to support high cell viabilities post-thawing. Controlled processes are used for cryopreservation, handling, and shipping to help minimize risk of adverse effects to the quality of the cells.
Made in USA by Stemedica for Thermo Fisher Scientific.
StemPro™ Neural Stem Cells help you to:
Immature, High Potency, Low Oxygen-Cultured NSCs
StemPro™ Neural Stem Cells are derived from human fetal brain from qualified, traceable donors. The cells are isolated, cultured, and expanded under Good Manufacturing Practice (GMP) manufacturing standards in a California-licensed facility using a proprietary Reduced Oxygen Tension manufacturing process. Manufacture of cells in a reduced oxygen tension environment results in higher yields of highly potent immature stem cells compared to cells expanded in normal oxygen culture conditions.
Safety Tested
StemPro™ Neural Stem Cells have been demonstrated to be non-toxic and non-tumorigenic in GLP animal studies performed according to FDA guidelines.
Large lots can be made available for those requiring assay standardization. Protocols using tested reagents are provided to help minimize your chance of assay failure using reagents and cells that have not been evaluated to work together. View our tested media and reagents.
High Post-Cryopreservation Viability
These StemPro™ Neural Stem Cells are supplied in CryoStor™ cryopreservation medium at a concentration of 5x106 viable cells/vial. CryoStor™ medium is animal origin-free and shown to support high cell viabilities post-thawing. Controlled processes are used for cryopreservation, handling, and shipping to help minimize risk of adverse effects to the quality of the cells.
Made in USA by Stemedica for Thermo Fisher Scientific.
Specifications
Specifications
| Content And Storage | Store in liquid nitrogen. |
| Age | Fetal |
| Cell Type | Neural Cells |
| Tissue | Cortex |
| Quantity | 1 x 106 cells |
| Tested For | Differentiation Potential, Pathogen, Sterility, Viability, Mycoplasma |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?