Learn More
Invitrogen™ STAT5 beta Recombinant Rabbit Monoclonal Antibody (24GB2110)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557740
 View more versions of this product
Product Code. 30276125
  • £488.00 / 100µL
Description
STAT5 beta Recombinant Monoclonal Antibody for Western Blot, Flow
The protein encoded by this gene is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein mediates the signal transduction triggered by various cell ligands, such as IL2, IL4, CSF1, and different growth hormones. It has been shown to be involved in diverse biological processes, such as TCR signaling, apoptosis, adult mammary gland development, and sexual dimorphism of liver gene expression. This gene was found to fuse to retinoic acid receptor-alpha (RARA) gene in a small subset of acute promyelocytic leukemias (APLL). The dysregulation of the signaling pathways mediated by this protein may be the cause of the APLL.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
STAT5 beta
Recombinant Monoclonal
0.53 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P42232, P51692, P52632
Stat5b
A synthesized peptide derived from human STAT5B (aa 500-787).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2110
Unconjugated
Stat5b
LOC101122961; mammary gland factor STAT5B; Signal transducer and activator of transcription 5a; signal transducer and activator of transcription 5B; STA5B; STAT; STAT5; STAT5B; transcription factor STAT5B
Rabbit
Affinity chromatography
RUO
20851, 25126, 6777
-20°C