missing translation for 'onlineSavingsMsg'
Learn More

Staphylococcus Aureus Enterotoxin B Antibody (S222), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody

£393.00

Specifications

Antigen Staphylococcus Aureus Enterotoxin B
Clone S222
Applications Immunoassay
Classification Monoclonal
Conjugate DyLight 594
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18576827
View Documents
Novus Biologicals
NB110-8272DL594
0.2 mL
£393.00
0.20mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Staphylococcus Aureus Enterotoxin B Monoclonal antibody specifically detects Staphylococcus Aureus Enterotoxin B in Bacteria samples. It is validated for Immunoassay, ELISA
Specifications

Specifications

Staphylococcus Aureus Enterotoxin B
Immunoassay
DyLight 594
Mouse
Bacteria
S. aureus
IgG1
Protein G purified
S222
Monoclonal
Purified
RUO
50mM Sodium Borate
Hybridoma clone has been derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with recombinant S. aureus enterotoxin B expressed in E. coli.
Primary
Store at 4°C in the dark.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.