missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Staphylococcus Aureus Enterotoxin B Antibody (S222), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-8272DL594
This item is not returnable.
View return policy
Description
Staphylococcus Aureus Enterotoxin B Monoclonal antibody specifically detects Staphylococcus Aureus Enterotoxin B in Bacteria samples. It is validated for Immunoassay, ELISA
Specifications
| Staphylococcus Aureus Enterotoxin B | |
| Monoclonal | |
| DyLight 594 | |
| S. aureus | |
| Hybridoma clone has been derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with recombinant S. aureus enterotoxin B expressed in E. coli. | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunoassay | |
| S222 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction