missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Glass TLC plates, SIL G/UV 254
- Pre-coated, ready-to-use plates
- Broad range of plate sizes, surface coatings, and backings
Absorbent (Silica 60)
- Specific surface (BET): 500 m2/g
- Mean pore size: 60 Å
- Specific pore volume: 0.75 mL/g
- Particle size: 5 to 17 μm
- Standard grade
Thickness
- Analytical plates: 0.25 mm
- Preparative plates: 0.5 mm and 1 mm
- 2 mm preparative layers use slightly coarser material
Indicators
- Manganese-activated zinc silicate with green fluorescence (short-wave UV, 254 nm)
- Special inorganic fluorescent pigment with blue fluorescence (long-wave UV, 366 nm)
Binders:
- Highly polymeric products
- Stable in almost all organic solvents
- Resistant to aggressive visualization reagents
Specifications
Specifications
| Backing Material | Glass |
| Material | Unmodified Silica Gel (SiOH) |
| Particle Size | 5 to 17 μm |
| Pore Size | 60 Å |
| Quantity | 200 Pc. |
| Type | Glass Plates Coated with Silica Gel |
| Fluorescent Indicator | No |
| For Use With (Application) | Steroids, Antibiotics, Amines, Amino acids, Hormones, Sweeteners, Aflatoxins, Vitamins, Normal Phase (NP), Alkaloids, Analgesics, Marihuana Compounds, Patulin, Standard TLC Applications |
| For Use With (Equipment) | Thin Layer Chromatography (TLC) |
| Includes | 200x TLC Glass Plates (5 x 10 cm) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?