missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Standard Probe for Dry Block Heaters

Product Code. 17551896 Shop All Fisherbrand Products
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17551896

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Standard probe for in-sample or in-block temperature control.

  • Encased in stainless steel sheath
  • Comes with 350 mm cable
TRUSTED_SUSTAINABILITY

Specifications

Material Stainless Steel
Type Temperature Probe
Includes 350 mm Cable
Length (Metric) 30 mm
Diameter (Metric) Base 3 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.