missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Scaffolding Tubes with End Plugs

Product Code. 11502672 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
250 mm
1000 mm
1500 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11502672 1500 mm Each
11592662 1000 mm Each
11582662 250 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11502672
Only null left
Add to basket
Add to basket

Provides for sturdy scaffolding construction in the laboratory. Fisherbrand™ Scaffolding Tubes with End Plugs are manufactured from durable stainless steel.

  • Approx. 13mm diameter
  • Available in a range of lengths
  • Fits with Fisherbrand scaffolding connectors and foot adapters

Specifications

Material Stainless Steel
Length (Metric) 1500 mm
Product Type Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.