missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Stainless Steel Collecting Pan with Outlet for Test Sieves

Collects resulting samples from test sieves. These stainless steel pans are compatible with multiple test sieves and sieve shakers, based upon the required diameter. Outlet allows draining of liquids during wet sieving.

Brand:  RETSCH 69.440.0070

Additional Details : Weight : 5.00000kg

 View more versions of this product

Product Code. 11934432

  • £765.00 / Each
Estimated Shipment: 30-07-2024
to see stock.
Add to basket

Description

Description

  • Stainless steel
  • Outlet allows draining to occur during separation or sorting
  • Compatible with multiple test sieves, based upon diameter
  • Excellent product quality due to extensive optical inspection
  • Maximum stability and optimum sealing when used in sieve stacks with O-ring
  • Clear and precise labeling of the sieves with full traceability based on individualized laser engraving
Specifications

Specifications

400 mm
Stainless Steel
70mm
Collecting Pan With Outlet
Test sieves; Analytical Sieve Shaker AS 200 Digital
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers