Learn More
Day Impex™ Stainless Steel Tempering Beaker
Product Code. 11827472
Change view
Click to view available options
Capacity (Metric):
600 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
11827472 600 mL Each
1 options
Product Code. 11827472 Supplier Day Impex™ Supplier No. 260121
Only null left
Add to basket
Add to basket
Day Impex™ Stainless Steel Tempering Beaker is a double walled beaker with ports for controlling the temperature inside the beaker.
  • Double walled beaker
  • Includes inlet and outlet ports for controlling temperature
  • Constructed of stainless steel
Specifications
Capacity (Metric) 600 mL
Material Stainless Steel
Type Beaker