Learn More
VACUUBRAND™ Stainless Steel Reducing Pieces With Small KF Flanges
Shop All Vacuubrand Products
Click to view available options
Diameter (Metric) Inside:
25/10 mm
25/16 mm
40/10 mm
40/16 mm
40/25 mm
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) Inside | 25/10 mm |
| For Use With | Solenoid-operated valves VV and VV C |
| Material | Stainless Steel |
| Quantity | 1 Each |
| Size | 40 mm |
| Type | Reducing Cross Piece |