Learn More
Bochem™ Stainless Steel Rectangle Test Tube Stands
Brand:  Bochem™ 8220
Code : A0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10535031
  • £38.60 / Each
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Specifications
Test Tube Rack
Tubes (20 mm)
12
2 x 6
Stainless Steel (18/10)
20 mm