Learn More
Endecotts™ Stainless Steel Receiver
Full height receivers for use with test sieves. Endecotts™ Stainless Steel Receivers are widely used with test sieves for all types of laboratory sampling and particle size analysis.
Brand:  Endecotts™ 100S/STREC
 View more versions of this product
Product Code. 12934225
  • £39.90 / Each
Estimated Shipment: 11-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Full height receivers for use with test sieves. Endecotts™ Stainless Steel Receivers are widely used with test sieves for all types of laboratory sampling and particle size analysis.
  • Stainless steel construction
  • For use with Stainless Steel Woven Wire Mesh Test Sieves
Specifications
Specifications
Receiver
100 mm
1 Ea.
Stainless Steel
For sieves