Learn More
Endecotts™ Stainless Steel Receiver
Product Code. p-8005592
Click to view available options
Diameter (Metric):
100 mm
200 mm
230 mm
300 mm
450 mm
Unit Size:
Each
Product Code. 12934225
Brand: Endecotts™ 100S/STREC
Only null left
Add to basket
Add to basket
Full height receivers for use with test sieves. Endecotts™ Stainless Steel Receivers are widely used with test sieves for all types of laboratory sampling and particle size analysis.
  • Stainless steel construction
  • For use with Stainless Steel Woven Wire Mesh Test Sieves
Specifications
Product Type Receiver
Material Stainless Steel
Diameter (Metric) 100 mm
For Use With (Application) For sieves
Packaging Quantity 1 Ea.