Learn More
Endecotts™ Stainless Steel Receiver
Full height receivers for use with test sieves. Endecotts™ Stainless Steel Receivers are widely used with test sieves for all types of laboratory sampling and particle size analysis.
Brand:  Endecotts™ 450S/STREC
Code : 03
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12825691
  • £124.00 / Each
Estimated Shipment: 01-07-2024
to see stock.
Add to basket
Description
Description
Full height receivers for use with test sieves. Endecotts™ Stainless Steel Receivers are widely used with test sieves for all types of laboratory sampling and particle size analysis.
  • Stainless steel construction
  • For use with Stainless Steel Woven Wire Mesh Test Sieves
Specifications
Specifications
Bottom Shelter for Screening
Stainless Steel
450 mm
1 Ea.