missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Stainless Steel Reagent Digger Spatula

Product Code. 11800100 Shop All Cole-Parmer Products
Click to view available options
Length (Metric):
165 mm
Unit Size:
Pack of 12
This item is not returnable. View return policy

Product Code. 11800100

Brand: Cole-Parmer™ 0101913

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Facilitates scooping and measuring of powdered, flaked, crystalline, or granular media

  • Especially useful for dislodging caked or crystallized reagents
  • Uniquely shaped stainless steel spatula fits reagent vessels with openings as small as 15⁄16 in. (2.4cm) dia.

Specifications

Quantity 12 Pk.
Shape Uniquel
Product Type Powder/Crystal/Grain Sampler
For Use With (Application) Useful For Dislodging Caked or Crystallized Reagents
Diameter (Metric) Outer 381mm
Material Stainless Steel
Length (Metric) 165 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.