missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Stainless Steel PTFE Coated Trulla Spatulas

Product Code. 11563512 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
175 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11563512 175 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11563512
Only null left
Add to basket
Edge
Add to basket

Ideal for Biology, Microbiology and Pharmaceutical Applications

  • Trulla Spatula PTFE coated
  • Supplied in standard packs of 1
  • Excellent heat resistance and great cryogenic stability
  • Applied as a self priming single coat PTFE
  • Coefficient of friction is generally in the range of 0.05 to 0.20

Specifications

Quantity 1 Ea.
Coating PTFE Xylan™
Product Type Trulla Spatula
For Use With (Application) Ideal for Biology, Microbiology and Pharmaceutical Applications
Temperature (Metric) Max -195°C to +260°C
Material Stainless Steel
Length (Metric) 175 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.