missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Stainless Steel Measuring Spoons

Product Code. 15366297 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
1.25 to 15 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15366297

Brand: Buerkle™ 53348000

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Set of 4. Measuring aid for fast and easy measuring from 1.25 to 15 mL. The volume is engraved on the handle of each measuring spoon.

  • Stainless steel AISI 304 (1.4301)
  • Volumes: 1.25 ml, 2.5 ml, 5 ml, 15 ml
  • Spoon high-gloss finished, handle matt polished
TRUSTED_SUSTAINABILITY

Specifications

Quantity 4 spoons
Capacity (Metric) 1.25 to 15 mL
Color Silver
For Use With (Application) Foodstuffs
Material Stainless Steel (V2A)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.