Learn More
Fisherbrand™ Stainless Steel Manual Decapper for clean room
Product Code. 11845722 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Unit Size:
Each
Size:
13 mm
20 mm
32 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size Size
11845722 Each -
11520535 Each 20 mm
15259968 Each 32 mm
11586034 Each 13 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11845722
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Allow easy removal of aluminum seals without breakage
  • Stainless Steel
  • Sterilizable for cleanroom applications
  • Manual
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Description 11mm Decapper
For Use With (Equipment) Aluminum Cap