missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Ladle Scoop with Flat Handle

Handle laboratory needs with the Bochem™ 18/10 Stainless Steel Scoop with Flat Handle utensil.

Brand:  Bochem™ 8745

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10759681

  • £101.00 / Each
Estimated Shipment: 21-06-2024
to see stock.
Add to basket

Description

Description

  • Flat Handle
  • Stainless Steel
  • Ladle-shaped scoop
Specifications

Specifications

1 Ea.
AISI 304
180mm
Flat
1400 mL
Ladle Scoop
Stainless Steel
350 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers