missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Stainless Steel Grinding Balls

Product Code. 10196541 Shop All Retsch Products
Change view
Click to view available options
For Use With (Equipment):
MM 200 and MM 301
Planetary Ball Mill PM 400
Reaction Vials
Diameter (Metric):
3 mm
4 mm
5 mm
7 mm
15 mm
40 mm
Unit Size:
Each
Pack of 200
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Diameter (Metric) unitSize
10196541 Reaction Vials 3 mm Each
10208591 Planetary Ball Mill PM 400 40 mm Each
10729954 MM 200 and MM 301 15 mm Each
10146641 Reaction Vials 5 mm Pack of 200
10054460 MM 200 and MM 301 7 mm Each
10539711 MM 200 and MM 301 5 mm Each
11758414 Reaction Vials 3 mm Pack of 200
11727664 Reaction Vials 4 mm Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10196541 Supplier RETSCH Supplier No. 22.455.0002
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed for use with Retsch™ mixer mills. Retsch™ Stainless Steel Grinding Balls are ideal for disruption of biological cells as well as for DNA/RNA extraction.

  • For use with MM 200 and MM 400 Mixer Mills

Specifications

Product Type Grinding Balls
Diameter (Metric) 3 mm
For Use With (Equipment) Reaction Vials
Material Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.