missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Grinding Balls for Geno/Grinder™ Homogenizers
Use grinding balls to grind samples in Geno/Grinder™ homogenizers.
£278.00 - £392.00
Product Code | Brand | Diameter (English) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) | Diameter (Metric) | Price | Quantity & Availability | ||||
12849055
|
Cole-Parmer™
2155 |
0.37 in. | 9.5 mm |
£278.00
Pack of 100 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||||
13591160
|
Cole-Parmer™
2156 |
0.43 in. | 1.1 cm |
£392.00
Pack of 100 |
Estimated Shipment: 20-11-2024
Log in to see stock. |
||||
Description
- Suitable for use with the Cole-Parmer™ Geno/Grinder™ homogenizers.
- Optimize performance of your Geno/Grinder™ homogenizers with a wide range of grinding ball options suitable for your application.
Specifications
Grinding Balls | |
Autoclavable |