Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class F2
Use with weighing instruments of high accuracy class II. Fisherbrand™ Class F2 Stainless Steel Calibration Weights are also intended for use in important commercial transactions, such as those involving precious metals and stones.
Brand:  Fisherbrand™
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15525119
  • £310.58 / 1 unit
Estimated Shipment: 21-06-2024
to see stock.
Add to basket
Description
Description
  • Class F2 stainless steel calibration weight
  • Weights intended for use in the verification or calibration of class M1 and possibly class M2 weights
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
F2
1 to 100 g
1 g to 100 g
Cylindrical