Learn More
Nickel Electro™ Stainless Steel Bucket with Handle
Product Code. 12882024 Shop All Nickel-Electro Products
Change view
Click to view available options
Unit Size:
Each
Product Code. Unit Size
12882024 Each
1 options
Product Code. 12882024 Supplier Nickel Electro™ Supplier No. BUCS0080
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Nickel Electro™ Stainless Steel Bucket with Handle provides good chemical resistance and can handle hot liquids.
  • 8 Litre Bucket
  • Stainless steel construction without graduations
  • Bright polished finish for easy cleaning and stainless steel wire handle
  • Good chemical resistance and can handle hot liquids
  • Stable in use
  • Can be autoclaved
Specifications
Product Type Bucket
Material Stainless Steel
Autoclavable Autoclavable
Handle Material Stainless steel
Includes Bright Polished finish
Quantity Ea.
Surface Type Bright Polished