Learn More
Bochem™ Stainless Steel Beakers With Spout and Handle
Product Code. 10430821
Change view
Click to view available options
:
Stainless Steel Beaker; Capacity: 5000 mL
Product Code.
10430821 Stainless Steel Beaker; Capacity: 5000 mL
1 options
Product Code. 10430821 Supplier Bochem™ Supplier No. 8536
Only null left
Add to basket
Add to basket
These Bochem™ 18/10 stainless steel beakers have a rim, spout and handle. They have a special contact base which make them ideal for use with hotplates and magnetic stirrers.
  • 18/10 stainless steel
  • Special-contact base for hotplates and magnetic stirrers
  • With pouring spout and handle
  • 5000 mL capacity
Specifications
Description Stainless Steel Beaker
Capacity (Metric) 5000 mL
Material Stainless Steel (18/10)
For Use With (Equipment) General Lab Use, Hotplates and Magnetic Stirrers
Includes Rim, Pouring Spout and Handle
Type Beaker