missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Stainless Steel Beaker

Bochem™ Stainless Steel Beaker with contact bottom for heating plates and magnetic stirrers.

Brand:  Bochem™ 8514

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10017800

  • £221.00 / Each
Estimated Shipment: 22-06-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Stainless Steel
  • For use with heating plates and magnetic stirrers
Specifications

Specifications

120mm
Stainless Steel (18/10)
With rim and spout
2000 mL
180mm
Videos
SDS
Documents
Certificates
Special Offers

Special Offers