Learn More
Bochem™ Stainless Steel Beakers
Product Code. 10781392
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
5000 mL
Includes:
Rim
Rim and Spout
Rim, Handle
Rim, Spout
Rim, Spout, Handle
Unit Size:
Each
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Includes unitSize
10781392 100 mL Rim, Spout, Handle Each
10586181 100 mL Rim, Handle Each
10498871 250 mL Rim, Spout, Handle Each
10201081 250 mL Rim Each
10142651 250 mL Rim, Handle Each
10663891 250 mL Rim, Spout Each
10288690 500 mL Rim, Spout, Handle Each
10017730 500 mL Rim Each
10603511 1000 mL Rim Each
10470061 1000 mL Rim, Handle Each
10218840 1000 mL Rim, Spout, Handle Each
10343191 1000 mL Rim and Spout Each
10566932 2000 mL Rim, Handle Each
10303051 2000 mL Rim Each
10627341 2000 mL Rim, Spout, Handle Each
10330731 3000 mL Rim, Spout, Handle Each
10363091 3000 mL Rim Each
10271361 3000 mL Rim, Spout Each
10729621 5000 mL Rim Each
10453501 5000 mL Rim, Spout Each
10244081 5000 mL Rim, Handle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Product Code. 10781392 Supplier Bochem™ Supplier No. 8530
Only null left
Add to basket
Add to basket
Highly resistant and durable stainless steel beaker. Bochem™ Stainless Steel Beakers include rims, spouts, and handles in various model combinations.
  • 18/10 stainless steel construction.
  • Resistant and durable.
  • Contact bottom for heating plates and magnetic stirrers.
Specifications
Capacity (Metric) 100 mL
Material Stainless Steel (18/10)
Includes Rim, Spout, Handle
Type Beaker