missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilac™ Stackable Pedestal Crate
Click to view available options
Color:
Gray
White
Unit Size:
Each
Description
- Storage bin with 4 feet made of food-grade plastic (HDPE)
- Solid stackable bin with great stability
- Slightly indented sides allow air flow once bins are stacked
- Professional quality, this food bin is designed for intensive use
Specifications
Specifications
| Type | Storage Bin |
| Material | HDPE |
| Color | Gray |
| Shape | Rectangular |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction