missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Stackable Pedestal Crate

Product Code. 30245680
Click to view available options
Color:
Gray
White
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30245680

Brand: Gilac™ G156050

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Storage crate with 4 feet made of food-grade plastic (HDPE). Solid stackable crate with great stability.

  • Storage bin with 4 feet made of food-grade plastic (HDPE)
  • Solid stackable bin with great stability
  • Slightly indented sides allow air flow once bins are stacked
  • Professional quality, this food bin is designed for intensive use

Specifications

Type Storage Bin
Material HDPE
Color Gray
Shape Rectangular
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.