missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilac™ Stackable Pedestal Bin
Description
- Storage bin with 4 feet made of food-grade plastic (HDPE)
- Solid stackable bin with great stability
- Slightly indented sides allow air flow once bins are stacked
- Professional quality, this food bin is designed for intensive use
Specifications
Specifications
| Type | Storage Bin |
| Material | HDPE |
| Color | Gray |
| Shape | Rectangular |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?