missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
£163.00 - £288.00
Specifications
| Antigen | ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 |
|---|---|
| Dilution | Western Blot 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18583594
|
Novus Biologicals
NB100-75632-0.025mg |
0.025 mg |
£288.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18551671
|
Novus Biologicals
NB100-75632-0.02mg |
0.02 mg |
£163.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 Polyclonal specifically detects ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 in Human samples. It is validated for Western Blot.Specifications
| ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Q6PZN5 | |
| 6489 | |
| The immunogen for this antibody was a sequence from 4a.a. to 18a.a. from ST8SIA1(a 356a.a. long protein) | |
| Affinity Purified | |
| Ganglioside GD3 |
| Western Blot 1:100-1:2000 | |
| Polyclonal | |
| Chicken | |
| Neuronal Stem Cell Markers, Stem Cell Markers | |
| PBS with No Preservative | |
| Alpha-2,8-sialyltransferase 8A, alpha-N-acetylneuraminide alpha-2,8-sialyltransferase, disialoganglioside (GD3) synthase, Ganglioside GD3 synthase, Ganglioside GT3 synthase, ganglioside-specific alpha-2,8-polysialyltransferase, GD3 synthase), GD3S, sialyltransferase 8 (alpha-N-acetylneuraminate: alpha-2,8-sialytransferase, GD3synthase) A, Sialyltransferase 8A, sialyltransferase 8A (alpha-N-acetylneuraminate: alpha-2,8-sialyltransferase, Sialytransferase St8Sia I, SIAT8A, SIAT8-A, SIAT8EC 2.4.99.8, ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 1, ST8SiaI | |
| ST8SIA1 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title