missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NB100-75632-0.02mg
This item is not returnable.
View return policy
Description
ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 Polyclonal specifically detects ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 in Human samples. It is validated for Western Blot.
Specifications
| ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 | |
| Polyclonal | |
| Western Blot 1:100-1:2000 | |
| Q6PZN5 | |
| ST8SIA1 | |
| The immunogen for this antibody was a sequence from 4a.a. to 18a.a. from ST8SIA1(a 356a.a. long protein) | |
| 0.02 mg | |
| Neuronal Stem Cell Markers, Stem Cell Markers | |
| 6489 | |
| Human |
| Western Blot | |
| Unconjugated | |
| PBS with No Preservative | |
| Alpha-2,8-sialyltransferase 8A, alpha-N-acetylneuraminide alpha-2,8-sialyltransferase, disialoganglioside (GD3) synthase, Ganglioside GD3 synthase, Ganglioside GT3 synthase, ganglioside-specific alpha-2,8-polysialyltransferase, GD3 synthase), GD3S, sialyltransferase 8 (alpha-N-acetylneuraminate: alpha-2,8-sialytransferase, GD3synthase) A, Sialyltransferase 8A, sialyltransferase 8A (alpha-N-acetylneuraminate: alpha-2,8-sialyltransferase, Sialytransferase St8Sia I, SIAT8A, SIAT8-A, SIAT8EC 2.4.99.8, ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 1, ST8SiaI | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Ganglioside GD3 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction