missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST6GAL1 Mouse, anti-Human, Clone: 1F11G5, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ST6GAL1 |
|---|---|
| Clone | 1F11G5 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18005575
|
Proteintech
68633-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18034505
|
Proteintech
68633-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ST6GAL1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ST6GAL1. This antibody recognizes human, pig antigen. The ST6GAL1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ST6GAL1 | |
| 1000 μg/mL | |
| Liquid | |
| Human, Pig | |
| Alpha 2,6 ST 1, B cell antigen CD75, CD75, Sialyltransferase 1, SIAT1, ST6Gal I, ST6GAL1, ST6GalI, ST6N | |
| Primary |
| 1F11G5 | |
| Western Blot, ELISA | |
| Mouse | |
| 6480 | |
| IgG2b | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title