Learn More
ST6GAL1 Mouse, anti-Human, Clone: 1F11G5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68633-1-IG-20UL
This item is not returnable.
View return policy
Description
The ST6GAL1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ST6GAL1. This antibody recognizes human, pig antigen. The ST6GAL1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ST6GAL1 | |
| 1F11G5 | |
| Alpha 2,6 ST 1, B cell antigen CD75, CD75, Sialyltransferase 1, SIAT1, ST6Gal I, ST6GAL1, ST6GalI, ST6N | |
| 20 μL | |
| 6480 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Pig | |
| 150 kDa | |
| IgG2b |
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido