missing translation for 'onlineSavingsMsg'
Learn More

ST3GAL4 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-95141-0.02ml

 View more versions of this product

Product Code. 18677902

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

ST3GAL4 Polyclonal antibody specifically detects ST3GAL4 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ST3GAL4
Polyclonal
Western Blot 1:500 - 1:2000, Immunohistochemistry-Paraffin
Alpha 2,3-sialyltransferase IV, Alpha 2,3-ST 4, Beta-galactoside alpha-2,3-sialyltransferase 4, CGS23FLJ46764, EC 2.4.99, EC 2.4.99.-, EC 2.4.99.9, FLJ11867, gal-beta-1,4-GalNAc-alpha-2,3-sialyltransferase, Gal-NAc6S, NANTA3alpha-3-N-acetylneuraminyltransferase, SAT3, SAT-3, Sialyltransferase 4C, sialyltransferase 4C (beta-galactosidase alpha-2,3-sialytransferase), sialyltransferase 4C (beta-galactoside alpha-2,3-sialytransferase), SIAT4-C, SIAT4CCMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 4, ST3 beta-galactoside alpha-2,3-sialyltransferase 4, ST3Gal IV, ST3GalA.2, ST3GalIV, ST-4, STZSIAT4
Recombinant fusion protein containing a sequence corresponding to amino acids 27-267 of human ST3GAL4 (NP_006269.1). REDSFYFPIPEKKEPCLQGEAESKASKLFGNYSRDQPIFLRLEDYFWVKTPSAYELPYGTKGSEDLLLRVLAITSSSIPKNIQSLRCRRCVVVGNGHRLRNSSLGDAINKYDVVIRLNNAPVAGYEGDVGSKTTMRLFYPESAHFDPKVENNPDTLLVLVAFKAMDFHWIETILSDKKRVRKGFWKQPPLIWDVNPKQIRILNPFFMEIAADKLLSLPMQQPRKIKQKPTTGLLAITLALH
0.02 mL
Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
6484
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.