missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST3GAL4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£156.00 - £364.00
Specifications
| Antigen | ST3GAL4 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Beschreibung
ST3GAL4 Polyclonal antibody specifically detects ST3GAL4 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Spezifikation
| ST3GAL4 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 6484 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| Alpha 2,3-sialyltransferase IV, Alpha 2,3-ST 4, Beta-galactoside alpha-2,3-sialyltransferase 4, CGS23FLJ46764, EC 2.4.99, EC 2.4.99.-, EC 2.4.99.9, FLJ11867, gal-beta-1,4-GalNAc-alpha-2,3-sialyltransferase, Gal-NAc6S, NANTA3alpha-3-N-acetylneuraminyltransferase, SAT3, SAT-3, Sialyltransferase 4C, sialyltransferase 4C (beta-galactosidase alpha-2,3-sialytransferase), sialyltransferase 4C (beta-galactoside alpha-2,3-sialytransferase), SIAT4-C, SIAT4CCMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 4, ST3 beta-galactoside alpha-2,3-sialyltransferase 4, ST3Gal IV, ST3GalA.2, ST3GalIV, ST-4, STZSIAT4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 27-267 of human ST3GAL4 (NP_006269.1). REDSFYFPIPEKKEPCLQGEAESKASKLFGNYSRDQPIFLRLEDYFWVKTPSAYELPYGTKGSEDLLLRVLAITSSSIPKNIQSLRCRRCVVVGNGHRLRNSSLGDAINKYDVVIRLNNAPVAGYEGDVGSKTTMRLFYPESAHFDPKVENNPDTLLVLVAFKAMDFHWIETILSDKKRVRKGFWKQPPLIWDVNPKQIRILNPFFMEIAADKLLSLPMQQPRKIKQKPTTGLLAITLALH | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts