missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
SSFA2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£159.00 - £364.00
Tekniske data
| Antigen | SSFA2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Beskrivelse
SSFA2 Polyclonal antibody specifically detects SSFA2 in Mouse samples. It is validated for Western BlotTekniske data
| SSFA2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.3), 50% glycerol | |
| 6744 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| Cleavage signal-1 protein, CS1, CS-1DKFZp313O1039, FLJ45996, KIAA1927, Ki-ras-induced actin-interacting protein, KRAPDKFZp779G0129, SPAG13, sperm specific antigen 2, sperm-specific antigen 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1000-1259 of human SSFA2 (NP_001123917.1). EVDQLQGLRNSVRMELQDLELQLEERLLGLEEQLRAVRMPSPFRSSALMGMCGSRSADNLSCPSPLNVMEPVTELMQEQSYLKSELGLGLGEMGFEIPPGESSESVFSQATSESSSVCSGPSHANRRTGVPSTASVGKSKTPLVARKKVFRASVALTPTAPSRTGSVQTPPDLESSEEVDAAEGAPEVVGPKSEVEEGHGKLPSMPAAEEMHKNVEQDELQQVIREIKESIVGEIRREIVSGLLAAVSSSKASNSKQDYH | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel