missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSFA2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93018-0.02ml
This item is not returnable.
View return policy
Description
SSFA2 Polyclonal antibody specifically detects SSFA2 in Mouse samples. It is validated for Western Blot
Specifications
| SSFA2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Cleavage signal-1 protein, CS1, CS-1DKFZp313O1039, FLJ45996, KIAA1927, Ki-ras-induced actin-interacting protein, KRAPDKFZp779G0129, SPAG13, sperm specific antigen 2, sperm-specific antigen 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1000-1259 of human SSFA2 (NP_001123917.1). EVDQLQGLRNSVRMELQDLELQLEERLLGLEEQLRAVRMPSPFRSSALMGMCGSRSADNLSCPSPLNVMEPVTELMQEQSYLKSELGLGLGEMGFEIPPGESSESVFSQATSESSSVCSGPSHANRRTGVPSTASVGKSKTPLVARKKVFRASVALTPTAPSRTGSVQTPPDLESSEEVDAAEGAPEVVGPKSEVEEGHGKLPSMPAAEEMHKNVEQDELQQVIREIKESIVGEIRREIVSGLLAAVSSSKASNSKQDYH | |
| 0.02 mL | |
| Cell Cycle and Replication | |
| 6744 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction