missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-5 Mouse, anti-Human, Unconjugated, Clone: 8e11, STEMCELL Technologies
Mouse Monoclonal Antibody
£202.50 - £531.90
Specifications
| Antigen | SSEA-5 |
|---|---|
| Clone | 8e11 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17158791
|
Stemcell Technologies
60063.1 |
50 μg |
£202.50
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17148791
|
Stemcell Technologies
60063 |
200 μg |
£531.90
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The 8e11 antibody reacts with stage-specific embryonic antigen-5 (SSEA-5), a terminal H type-1 glycan (Fuc1-2Galβ1-3GlcNAcβ1) expressed on the surface of cells in the inner cell mass of the human blastocyst during embryogenesis, and on undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells
- Expression of SSEA-5 is rapidly down-regulated upon cellular differentiation
- The 8e11 antibody can be used in conjunction with other pluripotency surface markers such as CD9 and CD90, or CD50 and CD200, to sort and remove undifferentiated teratoma-forming cells from incompletely differentiated stem cell cultures
Specifications
| SSEA-5 | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Human | |
| Primary |
| 8e11 | |
| Monoclonal | |
| Mouse | |
| IgG1 κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title