Learn More
SSEA-5 Mouse, anti-Human, Unconjugated, Clone: 8e11, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60063.1
This item is not returnable.
View return policy
Description
- The 8e11 antibody reacts with stage-specific embryonic antigen-5 (SSEA-5), a terminal H type-1 glycan (Fuc1-2Galβ1-3GlcNAcβ1) expressed on the surface of cells in the inner cell mass of the human blastocyst during embryogenesis, and on undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells
- Expression of SSEA-5 is rapidly down-regulated upon cellular differentiation
- The 8e11 antibody can be used in conjunction with other pluripotency surface markers such as CD9 and CD90, or CD50 and CD200, to sort and remove undifferentiated teratoma-forming cells from incompletely differentiated stem cell cultures
Specifications
| SSEA-5 | |
| Monoclonal | |
| Unconjugated | |
| 50 μg | |
| Human | |
| Pluripotent Stem Cells |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 8e11 | |
| Mouse | |
| Primary | |
| IgG1 κ |