missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-1 (CD15) Mouse, anti-Mouse, FITC, Clone: MC-480, STEMCELL Technologies
Mouse Monoclonal Antibody
£252.45 - £561.60
Specifications
| Antigen | SSEA-1 (CD15) |
|---|---|
| Clone | MC-480 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17118781
|
Stemcell Technologies
60060FI.1 |
25 Tests |
£252.45
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17108781
|
Stemcell Technologies
60060FI |
100 Tests |
£561.60
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The MC-480 antibody reacts with a terminal carbohydrate epitope, stage-specific embryonic antigen-1 (SSEA-1), which is expressed on a large-molecular-mass (> 200 kDa) glycoprotein on the surface of early mouse embryos, mouse embryonal carcinoma (EC), embryonic stem (ES) cells and mouse and human embryonic germ (EG) cells
- SSEA-1 is not expressed on undifferentiated human EC, ES or induced pluripotent stem (iPS) cells, or rhesus monkey ES cell lines
- It has been reported that SSEA-1 has roles in cell adhesion and migration, and regulation of cell differentiation
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ HLA Chimerism Whole Blood CD15 Positive Selection Kit (Catalog #17881; partial blocking may be observed), and for labeling human ES and iPS cells grown in TeSR™-E8™ (Catalog #05940), mTeSR™1 (Catalog #85850) and TeSR™2 (Catalog #05860)
Specifications
| SSEA-1 (CD15) | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| FITC | |
| Human, Rat, Mouse | |
| Primary |
| MC-480 | |
| Monoclonal | |
| Mouse | |
| IgM κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title