Learn More
SSEA-1 (CD15) Mouse, anti-Mouse, FITC, Clone: MC-480, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60060FI
This item is not returnable.
View return policy
Description
- The MC-480 antibody reacts with a terminal carbohydrate epitope, stage-specific embryonic antigen-1 (SSEA-1), which is expressed on a large-molecular-mass (> 200 kDa) glycoprotein on the surface of early mouse embryos, mouse embryonal carcinoma (EC), embryonic stem (ES) cells and mouse and human embryonic germ (EG) cells
- SSEA-1 is not expressed on undifferentiated human EC, ES or induced pluripotent stem (iPS) cells, or rhesus monkey ES cell lines
- It has been reported that SSEA-1 has roles in cell adhesion and migration, and regulation of cell differentiation
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ HLA Chimerism Whole Blood CD15 Positive Selection Kit (Catalog #17881; partial blocking may be observed), and for labeling human ES and iPS cells grown in TeSR™-E8™ (Catalog #05940), mTeSR™1 (Catalog #85850) and TeSR™2 (Catalog #05860)
Specifications
| SSEA-1 (CD15) | |
| Monoclonal | |
| FITC | |
| 100 Tests | |
| Human, Rat, Mouse | |
| Pluripotent Stem Cells |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| MC-480 | |
| Mouse | |
| Primary | |
| IgM κ |