missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-1 Antibody (MC-480), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
£186.00 - £412.00
Specifications
| Antigen | SSEA-1 |
|---|---|
| Clone | MC-480 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 ug/ml, Simple Western 1:10, Flow Cytometry 2.5 ug/million cells, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18009223
|
Novus Biologicals
NB100-1831SS |
0.025 mL |
£186.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18775853
|
Novus Biologicals
NB100-1831 |
0.1 mL |
£412.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SSEA-1 Monoclonal specifically detects SSEA-1 in Human, Mouse, Rat, Chimpanzee samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| SSEA-1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells | |
| PBS with 0.02% Sodium Azide | |
| 2526 | |
| F9 tetracarcinoma stem cells (X-irradiated). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MC-480 | |
| Western Blot 2 ug/ml, Simple Western 1:10, Flow Cytometry 2.5 ug/million cells, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Chimpanzee | |
| alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1 | |
| FUT4 | |
| IgM | |
| IgM purified | |
| Human and mouse. This antibody recognizes SSEA1 in mouse EC and ES cells but not human, but SSEA1 is expressed by some human differentiated cell types (ie: trophoblasts). Reactivity with Rat is reported by Kim et al. 2007 (PMID 17548530). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title